<iframe src="https://www.googletagmanager.com/ns.html?id=GTM-TXMJ2GG>m_auth=>m_preview=>m_cookies_win=x" height="0" width="0" style="display:none;visibility:hidden" id="tag-manager"></iframe>
Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Anthranilic acid
Synonym(s):
Anthranilic acid, 2-AA, 2-Aminobenzoic acid
Linear Formula:
2-(H
2
N)C
6
H
4
CO
2
H
CAS Number:
118-92-3
Molecular Weight:
137.14
EC Number:
204-287-5
Products
Product Category
organic acids (2)
acids (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
8.20112
Anthranilic acid,
for synthesis
Expand
View Pricing
A89855
Anthranilic acid,
reagent grade, ≥98%
Expand
View Pricing
10680
Anthranilic acid,
puriss. p.a., ≥99.5% (T)
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Español - ES
Learn More