Key Documents
PS641
Oxydemeton-methyl
analytical standard
Synonym(s):
Demeton-S-methyl sulfoxide, S-[2-(Ethylsulfinyl)ethyl] O,O-dimethyl phosphorothioate, Methyl oxydemeton
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C6H15O4PS2
CAS Number:
Molecular Weight:
246.28
EC Number:
UNSPSC Code:
41116107
grade
analytical standard
packaging
ampule of 50 mg
manufacturer/tradename
Chem Service, Inc. PS-641
application(s)
agriculture
environmental
storage temp.
2-8°C
InChI
1S/C6H15O4PS2/c1-4-13(8)6-5-12-11(7,9-2)10-3/h4-6H2,1-3H3
InChI key
PMCVMORKVPSKHZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Oxydemeton-methyl is an organophosphorus pesticide primarily used for the control of pests in a wide variety of crops. Its mode of action involves the inhibition of cholinesterase enzyme activity.
Application
Oxydemeton-methyl may be used as a reference standard for the determination of oxydemeton-methyl in aqueous samples by direct injection using liquid chromatography coupled with tandem mass spectrometry.
Footnote
This product has a 6 month shelf life from date of manufacture. We provide the newest lot available.
pictograms
Skull and crossbonesEnvironment
signalword
Danger
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
H Thiermann et al.
Human & experimental toxicology, 16(8), 473-480 (1997-08-01)
1 The effectiveness of oxime therapy in organophosphate poisoning is still a matter of debate. It appears, however, that the often cited ineffectiveness of oximes may be due to inappropriate dosing. By virtue of in vitro findings and theoretical considerations
D R Lenselink et al.
Teratology, 48(3), 207-211 (1993-09-01)
The teratogenic potential of oxydemeton-methyl (ODM) was investigated using a stage specific localized topical method of application to the stage 12 chick embryo. A dose of 0.01, 0.05, 0.10, 0.50, 1.0, or 2.0 mg/embryo was applied to the vitelline membrane
S Kleinschmidt et al.
Anasthesiologie, Intensivmedizin, Notfallmedizin, Schmerztherapie : AINS, 29(4), 248-251 (1994-06-01)
We report on a 52-year old woman who had taken an unknown dose of the organophosphate insecticide Metasystox R with suicidal intention. Ingestion was followed by cardio-respiratory failure. After successful cardio-pulmonary resuscitation, main therapeutic principles were the immediate application of
Determination of polar organophosphorus pesticides in aqueous samples by direct injection using liquid chromatography-tandem mass spectrometry.
Ingelse BA, et al.
Journal of Chromatography A, 918(1), 67-78 (2001)
D K Kansal et al.
Indian journal of physiology and pharmacology, 44(1), 57-63 (2000-08-05)
Oxydemeton-methyl, an organophosphate insecticide and acaricide produced decrease in the exploratory behaviour and prolongation of barbitone sodium-induced hypnosis after intermittent aerosol spray inhalational exposure for 1 h in rats compared to the saline control group. Further, CD50 +/- S.E.M. value