Key Documents
48156
Supelco
1,2,3,5-Tetrachlorobenzene solution
certified reference material, 2000 μg/mL in methanol
Sign Into View Organizational & Contract Pricing
About This Item
CAS Number:
Beilstein/REAXYS Number:
1618864
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
feature
standard type calibration
packaging
ampule of 1 mL
concentration
2000 μg/mL in methanol
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
single component solution
storage temp.
2-8°C
SMILES string
Clc1cc(Cl)c(Cl)c(Cl)c1
InChI
1S/C6H2Cl4/c7-3-1-4(8)6(10)5(9)2-3/h1-2H
InChI key
QZYNWJQFTJXIRN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Other Notes
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Legal Information
TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
49.5 °F - Solvent
flash_point_c
9.7 °C - Solvent
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Peter Kämpfer et al.
International journal of systematic and evolutionary microbiology, 54(Pt 3), 749-751 (2004-05-15)
A Gram-positive, rod-shaped, non-spore-forming bacterium (B5(T)) was isolated from an enrichment culture that contained 1,2,3,5-tetrachlorobenzene as the sole source of carbon. On the basis of 16S rRNA gene sequence similarity studies, strain B5(T) was shown to belong to the family
Mei Sun et al.
Environmental science & technology, 44(21), 8209-8215 (2010-10-01)
Sediment caps that degrade contaminants can improve their ability to contain contaminants relative to sand and sorbent-amended caps, but few methods to enhance contaminant degradation in sediment caps are available. The objective of this study was to determine if, carbon
W De Wolf et al.
Comparative biochemistry and physiology. C, Comparative pharmacology and toxicology, 100(1-2), 55-57 (1991-01-01)
1. Seven chlorinated anilines and one chlorinated benzene were tested for their ability to bioconcentrate in guppies (Poecilia reticulata) under different experimental conditions. 2. Interactions between compounds in a mixture influence the bioconcentration of some chlorinated anilines. These interactions result
B Z Fathepure et al.
Applied and environmental microbiology, 54(2), 327-330 (1988-02-01)
Hexachlorobenzene was dechlorinated to tri- and dichlorobenzenes in anaerobic sewage sludge. The complete biotransformation of 190 microM hexachlorobenzene (approximately 50 ppm) occurred within 3 weeks. The calculated rate of hexachlorobenzene dechlorination was 13.6 mumol liter-1 day-1. Hexachlorobenzene was dechlorinated via
M Parrini et al.
Bollettino della Societa italiana di biologia sperimentale, 66(7), 709-716 (1990-07-01)
The mutagenicity of halogenated benzenes, including three isomers of tri- and tetrachlorobenzenes (TCB, TeCB) was studied on male Swiss CD1 mice by MN test. The data presented show that all the halogenated benzenes tested were found to be clastogenic apart
Questions
Reviews
No rating value
Active Filters