Key Documents
Z378267
Drosophila Cells in Culture
Sign Into View Organizational & Contract Pricing
About This Item
ISBN-10:
0-12-229460-2
ISBN-13:
978-0-12-229460-0
UNSPSC Code:
55101509
publication info
G. Echalier, Academic Press, 1997, 702 pp., hard cover
manufacturer/tradename
Elsevier
General description
A pioneer in the use of Drosophila as an experimental model for developmental biology summarizes 30 years of experience in handling Drosophila cells in culture. The author emphasizes gene transfer methods, cell responses to heat shock, hormonal regulation of expression, and expression of transposable genetic elements.
Table of Contents
Composition of the Body Fluid of Drosophila and the Design of Culture Media for Drosophila Cells.
Primary Cell Cultures of Drosophila Cells.
Drosophila Continuous Cell Lines.
Karyotype and Cell Cycle.
Biology and Biochemistry of Cultured Cell Lines: Nucleic Acids.
Biology and Biochemistry of Cultured Cell Lines: Proteins.
Experimental Models of Gene Regulation: Heat-Shock Response of Drosophila Cells.
Experimental Models of Gene Regulation: Cell Responses to Hormones.
Gene Transfer Into Cultured Drosophila Cells.
Transposons.
Drosophila Viruses and Other Infections of Cultured Cells.
Chapter References.
Appendices.
Bibliography.
Subject Index.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.