Key Documents
Z1000
Sigma-Aldrich
Zinc bromide
Synonym(s):
Zinc dibromide
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
ZnBr2
CAS Number:
Molecular Weight:
225.20
EC Number:
MDL number:
UNSPSC Code:
12352103
mp
394 °C (lit.)
SMILES string
Br[Zn]Br
InChI
1S/2BrH.Zn/h2*1H;/q;;+2/p-2
InChI key
VNDYJBBGRKZCSX-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)