Key Documents
V4503
Sigma-Aldrich
[des-Gly9-β-Mercapto-β,β-cyclopentamethylenepropionyl1,O-Et-Tyr2,Val4,Arg8]-Vasopressin
≥97% (HPLC)
Synonym(s):
(d(CH2)51, Tyr(Et)2, Val4, Arg8, des-Gly-NH29)-Vasopressin
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C51H74N12O10S2
CAS Number:
Molecular Weight:
1079.34
MDL number:
UNSPSC Code:
12352200
assay
≥97% (HPLC)
storage temp.
−20°C
SMILES string
CCOc1ccc(C[C@@H]2NC(=O)CC3(CCCCC3)SSC[C@H](NC(=O)[C@H](CC(N)=O)NC(=O)[C@@H](NC(=O)[C@H](Cc4ccccc4)NC2=O)C(C)C)C(=O)N5CCC[C@H]5C(=O)N[C@@H](CCCNC(N)=N)C(N)=O)cc1
Amino Acid Sequence
(1-Mercaptocyclohexyl)acetyl-Tyr(O-Ethyl)-Phe-Val-Asn-Cys-Pro-Arg-NH2 [Disulfide Bridge: 1-6]
Biochem/physiol Actions
Potent V1 (vasopressor) and V2 (antidiuretic) vasopressin receptor antagonist.
pictograms
Exclamation mark
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Inhalation
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Determination of a disulfide-containing octapeptide antagonist of vasopressin in human plasma utilizing high-performance liquid chromatography with fluorescence detection.
G R Rhodes et al.
Journal of chromatography, 488(2), 456-462 (1989-03-24)
C R Albrightson-Winslow et al.
The Journal of pharmacology and experimental therapeutics, 249(2), 366-371 (1989-05-01)
The vasopressin (AVP) antagonist, SK&F 101926 (beta-mercapto-beta beta-cyclopentamethylene propionic acid1, D-Tyr(Et)2, Phe3, Val4, Asn5, Cys6, Pro7, Arg8-NH2), is an antidiuretic antagonist in rats and squirrel monkeys in vivo. In rat, dog, pig, squirrel monkey and human in vitro studies, SK&F
N Aiyar et al.
The Journal of pharmacology and experimental therapeutics, 242(1), 217-221 (1987-07-01)
We have previously reported that 8-arginine vasopressin (AVP) stimulates phosphatidylinositol turnover and Ca++ mobilization in rat aortic smooth muscle cells (A10). In the present study, N-ethylmaleimide (NEM) was used to further characterize the putative guanine nucleotide binding protein that transduces
F L Stassen et al.
Molecular pharmacology, 31(3), 267-272 (1987-03-01)
We report the vasopressin receptor-binding properties of [3H-Phe]-desGlyd(CH2)5D-Tyr(Et)VAVP, [3H]-SK&F 101926, the first radiolabeled vasopressin receptor antagonist. We chose to radiolabel SK&F 101926 because this vasopressin analog is a potent antagonist of vascular V1 and renal V2 vasopressin receptors in all
L B Kinter et al.
The American journal of physiology, 258(3 Pt 2), R763-R769 (1990-03-01)
Physiological regulation of receptor-effector pathways is recognized as a significant factor determining target organ selectivity and sensitivity in several hormonal systems. Whether or not physiological regulation of the renal vasopressin (V2) receptor-effector pathway participates in the control of body fluid