Key Documents
V2125
Sigma-Aldrich
Val-Val
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C10H20N2O3
CAS Number:
Molecular Weight:
216.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
storage temp.
−20°C
SMILES string
CC(C)C(N)C(=O)NC(C(C)C)C(O)=O
Looking for similar products? Visit Product Comparison Guide
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Uptake of L-valyl-L-valine and glycylsarcosine by hamster jejunum in vitro.
D Burston et al.
Clinical science (London, England : 1979), 62(6), 617-626 (1982-06-01)
[Effects of L-valyl-valine anhydride on the blood-perfused canine sinoatrial node and papillary muscle].
Y X Wang et al.
Zhongguo yao li xue bao = Acta pharmacologica Sinica, 7(5), 435-438 (1986-09-01)
D T Thwaites et al.
Biochemical and biophysical research communications, 194(1), 432-438 (1993-07-15)
Direct demonstration of intact peptide transport by the intestinal H+/dipeptide carrier is limited both by luminal/cytosolic hydrolysis and the availability of suitable radiolabelled substrates. Perfusion of Val-Val (20mM) at the apical surface of human intestinal epithelial (Caco-2) cell monolayers resulted
J Li et al.
Journal of drug targeting, 5(5), 317-327 (1998-10-15)
The objective of this study was to elucidate the structural features of the stereoisomers of Val-Val and Val-Val-Val that afford optimal binding affinity for the apical oligopeptide transporter in human intestinal Caco-2 cells. Three-dimensional conformations of cephalexin and Val stereoisomers
K Tamura et al.
Pharmaceutical research, 13(8), 1213-1218 (1996-08-01)
The purpose of this study was to determine whether the binding of the diastereomers of Val-Val to the apical oligopeptide transporter(s) could be correlated with their cellular uptake and transepithelial transport. The Caco-2 cell culture system was used for all