Key Documents
U1756
Sigma-Aldrich
Uridine 5′-diphosphoglucose-(glucose-1-3H)
aqueous ethanol solution
Synonym(s):
UDP-Glucose, UDP-glc, UDPG
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C15H22N2O17P2
CAS Number:
Molecular Weight:
564.29
MDL number:
UNSPSC Code:
12352200
form
aqueous ethanol solution
extent of labeling
2-12 Ci/mmol
shipped in
dry ice
storage temp.
−20°C
Physical form
Ethanol: water (70:30) solution
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.