Key Documents
T9267
Sigma-Aldrich
Glycerol triarachidate
Sigma Grade, ~99%
Synonym(s):
1,2,3-Triarachinoylglycerol, 1,2,3-Trieicosanoylglycerol, Glycerol triarachinate, Triarachin
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
[CH3(CH2)18COOCH2]2CHOCO(CH2)18CH3
CAS Number:
Molecular Weight:
975.64
Beilstein/REAXYS Number:
1811420
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
grade
Sigma Grade
assay
~99%
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCCCC
Looking for similar products? Visit Product Comparison Guide
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)