T9165
ψ-Tectorigenin
≥98%
Synonym(s):
5,7-Dihydroxy-3-(4-hydroxyphenyl)-8-methoxy-4H-1-benzopyran-4-one
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H12O6
CAS Number:
Molecular Weight:
300.26
MDL number:
UNSPSC Code:
51101500
PubChem Substance ID:
assay
≥98%
mode of action
enzyme | inhibits
storage temp.
2-8°C
SMILES string
COc1c(O)cc(O)c2C(=O)C(=COc12)c3ccc(O)cc3
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Imoto, M., et al.
J. Cell Pharmacol.,, 2, 40-40 (1991)
[Biological activity of oncogene function inhibitors isolated from microorganisms].
K Umezawa
Seikagaku. The Journal of Japanese Biochemical Society, 64(3), 160-171 (1992-03-01)
Inhibition of epidermal growth factor-induced activation of phospholipase C by psi-tectorigenin.
M Imoto et al.
The Journal of antibiotics, 44(8), 915-917 (1991-08-01)
[Inhibitors of growth factor functions, possible antineoplastic agents].
K Umezaza et al.
Tanpakushitsu kakusan koso. Protein, nucleic acid, enzyme, 36(7), 1429-1435 (1991-05-01)
M Imoto et al.
FEBS letters, 230(1-2), 43-46 (1988-03-28)
Psi-tectorigenin, an isoflavonoid, was isolated from a culture filtrate of actinomycetes as an inhibitor of epidermal growth factor-induced phosphatidylinositol turnover in cultured A431 cells. It inhibited phosphatidylinositol turnover with an IC50 of about 1 microgram/ml; thus, its inhibitory activity was