Key Documents
T6550
Sigma-Aldrich
2,9,16,23-Tetraphenoxy-29H,31H-phthalocyanine
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C56H34N8O4
CAS Number:
Molecular Weight:
882.92
MDL number:
UNSPSC Code:
12171500
composition
Dye content, ~98%
mp
195 °C (dec.) (lit.)
SMILES string
O(c1ccccc1)c2ccc3c4nc(nc5[nH]c(nc6nc(nc7[nH]c(n4)c8cc(Oc9ccccc9)ccc78)c%10cc(Oc%11ccccc%11)ccc6%10)c%12cc(Oc%13ccccc%13)ccc5%12)c3c2
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)