Key Documents
T6256
Sigma-Aldrich
N-Trifluoroacetyl-L-valine methyl ester
~98% (GC)
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C8H12F3NO3
CAS Number:
Molecular Weight:
227.18
MDL number:
UNSPSC Code:
12352200
assay
~98% (GC)
storage temp.
2-8°C
SMILES string
COC(=O)C(NC(=O)C(F)(F)F)C(C)C
Choose from one of the most recent versions:
Certificates of Analysis (COA)