Key Documents
T5193
Sigma-Aldrich
Tyrphostin AG 698
≥98%
Synonym(s):
N-(2-Phenylethyl)-3,4-dihydroxybenzylidenecyanoacetamide, Tyrphostin B52
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C18H16N2O3
Molecular Weight:
308.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
assay
≥98%
potency
IC50 (930 nanomolar for inhibition of EGFR-kinase activity.)
solubility
DMSO or ethanol: soluble
storage temp.
−20°C
SMILES string
Oc1ccc(cc1O)\C=C(/C#N)C(=O)NCCc2ccccc2
Gene Information
Biochem/physiol Actions
EGFR protein tyrosine kinase inhibitor.
Preparation Note
Tyrphostin AG 698 is soluble in DMSO and ethanol.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)