Key Documents
T4761
Sigma-Aldrich
Thymolphthalein monophosphate 2-amino-2-methyl-1,3-propanediol salt
90-95% (TLC)
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
C28H31O7P · (C4H11NO2)2
CAS Number:
Molecular Weight:
720.79
EC Number:
MDL number:
UNSPSC Code:
12352204
assay
90-95% (TLC)
storage temp.
−20°C
Suitability
Suitable for use as a substrate for alkaline phosphatase
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)