T4757
Glyceryl trinervonate
~98% (GC)
Synonym(s):
1,2,3-Tri(cis-15-tetracosenoyl)glycerol, Trinervonin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C75H140O6
CAS Number:
Molecular Weight:
1137.91
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
assay
~98% (GC)
storage temp.
−20°C
SMILES string
CCCCCCCC\C=C/CCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCC\C=C/CCCCCCCC)OC(=O)CCCCCCCCCCCCC\C=C/CCCCCCCC
Packaging
Sealed ampule.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)