Key Documents
T4149
Sigma-Aldrich
Glyceryl tripetroselinate
~99%
Synonym(s):
1,2,3-Tri(cis-6-octadecenoyl)glycerol, Tripetroselinin
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C57H104O6
CAS Number:
Molecular Weight:
885.43
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
assay
~99%
storage temp.
−20°C
SMILES string
CCCCCCCCCCC\C=C/CCCCC(=O)OCC(COC(=O)CCCC\C=C/CCCCCCCCCCC)OC(=O)CCCC\C=C/CCCCCCCCCCC
Looking for similar products? Visit Product Comparison Guide
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)