Key Documents
T1754
Sigma-Aldrich
Trp-Gly
≥97% (TLC)
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C13H15N3O3
CAS Number:
Molecular Weight:
261.28
MDL number:
UNSPSC Code:
12352200
assay
≥97% (TLC)
storage temp.
−20°C
SMILES string
NC(Cc1c[nH]c2ccccc12)C(=O)NCC(O)=O
Looking for similar products? Visit Product Comparison Guide
Other Notes
Dipeptide found in human pituitary gland.
Choose from one of the most recent versions:
Certificates of Analysis (COA)