Key Documents
T0932
Sigma-Aldrich
Testosterone-[1,2,6,7-3H(N)]
≥95% (Radiochemical Purity), ethanol solution
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C19H28O2
CAS Number:
Molecular Weight:
288.42
MDL number:
UNSPSC Code:
12352200
assay
≥95% (Radiochemical Purity)
form
ethanol solution
drug control
regulated under CDSA - not available from Sigma-Aldrich Canada
extent of labeling
85-105 Ci/mmol
mass shift
M+16
storage temp.
−20°C
SMILES string
[3H]C1([3H])[C@H]2[C@@H]3CC[C@H](O)[C@@]3(C)CC[C@@H]2[C@]4(C)C(=CC(=O)C([3H])([3H])C4([3H])[3H])C1([3H])[3H]
Packaging
serum bottle
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.