Key Documents
S8665
Sigma-Aldrich
Ser-Ala-Lys-Leu-Cys-Pro-Gly-Gly-Asn-Cys-Val [Disulfide bridge: 5−10]
≥97% (HPLC)
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C42H71N13O14S2
CAS Number:
Molecular Weight:
1046.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
assay
≥97% (HPLC)
UniProt accession no.
storage temp.
−20°C
SMILES string
CC(C)CC(NC(=O)C(CCCCN)NC(=O)C(C)NC(=O)C(N)CO)C(=O)NC1CSSCC(NC(=O)C(CC(N)=O)NC(=O)CNC(=O)CNC(=O)C2CCCN2C1=O)C(=O)NC(C(C)C)C(O)=O
Gene Information
mouse ... Npy(109648)
Biochem/physiol Actions
Sperm-activating peptide of sea urchin
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)