Key Documents
S4814
Sigma-Aldrich
N-Succinyl-Gly-Gly-Phe-4-methoxy-2-naphthylamide
≥98% (TLC), solid
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C28H30N4O7
Molecular Weight:
534.56
MDL number:
UNSPSC Code:
12352200
assay
≥98% (TLC)
form
solid
storage temp.
−20°C
Biochem/physiol Actions
May be used as a cathepsin G substrate
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.