S3629
Succinonitrile
Synonym(s):
1,2-Dicyanoethane, Butanedinitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NCCH2CH2CN
CAS Number:
Molecular Weight:
80.09
Beilstein/REAXYS Number:
1098380
EC Number:
MDL number:
UNSPSC Code:
12352100
Recommended Products
bp
265-267 °C (lit.)
mp
50-54 °C (lit.)
density
0.985 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
N#CCCC#N
InChI
1S/C4H4N2/c5-3-1-2-4-6/h1-2H2
InChI key
IAHFWCOBPZCAEA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)