Key Documents
S2907
Sigma-Aldrich
Salicylate Hydroxylase from Pseudomonas sp.
lyophilized powder, 0.5-3.0 units/mg protein
Synonym(s):
Salicylate 1-monooxygenase, Salicylate, NADH:oxygen oxidoreductase (1-hydroxylating, decarboxylating)
Sign Into View Organizational & Contract Pricing
About This Item
CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
form
lyophilized powder
specific activity
0.5-3.0 units/mg protein
composition
Protein, 35-65% biuret
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Unit Definition
One unit will convert 1.0 μmole of salicylate and β-NADH to catechol and β-NAD per min at pH 7.6 at 30 °C.
Physical form
Lyophilized powder containing Tris phosphate and FAD
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Thimmaraju Rudrappa et al.
Planta, 226(2), 283-297 (2007-06-08)
Bacillus subtilis, a plant growth promoting rhizobacteria (PGPR), induces growth response and protection against pathogenic organisms through colonization and biofilm formation on the Arabidopsis thaliana root surface. In the current investigation, we utilized various Arabidopsis defense pathway mutants in a
Yves Jouanneau et al.
Applied and environmental microbiology, 73(23), 7515-7521 (2007-10-02)
In the bacterial degradation of polycyclic aromatic hydrocarbons (PAHs), salicylate hydroxylases catalyze essential reactions at the junction between the so-called upper and lower catabolic pathways. Unlike the salicylate 1-hydroxylase from pseudomonads, which is a well-characterized flavoprotein, the enzyme found in
Josefa M Alamillo et al.
The Plant journal : for cell and molecular biology, 48(2), 217-227 (2006-10-05)
Plum pox virus (PPV) is able to replicate in inoculated leaves of Nicotiana tabacum, but is defective in systemic movement in this host. However, PPV produces a systemic infection in transgenic tobacco expressing the silencing suppressor P1/HC-Pro from tobacco etch
Beatriz Cámara et al.
Journal of bacteriology, 189(5), 1664-1674 (2006-12-19)
Pseudomonas sp. strain MT1 has recently been reported to degrade 4- and 5-chlorosalicylate by a pathway assumed to consist of a patchwork of reactions comprising enzymes of the 3-oxoadipate pathway. Genes encoding the initial steps in the degradation of salicylate
Shunji Ueno et al.
Free radical research, 41(11), 1246-1252 (2007-10-02)
Hydroxyl radical (*OH) generation in the kidney of mice treated with ferric nitrilotriacetate (Fe-NTA) or potassium bromate (KBrO3) in vivo was estimated by the salicylate hydroxylation method, using the optimal experimental conditions we recently reported. Induction of DNA lesions and