Key Documents
S2190
Sigma-Aldrich
Anti-STAT1α antibody produced in rabbit
affinity isolated antibody, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
About This Item
MDL number:
UNSPSC Code:
12352203
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
clone
polyclonal
form
buffered aqueous solution
species reactivity
mouse
should not react with
human
technique(s)
western blot: 0.1-0.5 μg/mL using 3T3-LI, NSF 60 or CTLL whole cell lysates
shipped in
dry ice
storage temp.
−70°C
Gene Information
mouse ... Stat1(20846)
Immunogen
mouse STAT1α
Physical form
Solution in phosphate buffered saline, pH 7.3, with no preservatives added.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
L B Ivashkiv
Reviews in immunogenetics, 2(2), 220-230 (2001-03-22)
Many, if not most, cytokines important for immune responses utilize the Jak-STAT signaling pathway. Jaks are receptor-associated protein tyrosine kinases, and STATs are latent cytoplasmic transcription factors that are activated by tyrosine phosphorylation. STATs play critical, nonredundant roles in mediating