Key Documents
S2011
Sigma-Aldrich
[Met-OMe11]-Substance P
≥97% (HPLC)
Synonym(s):
Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-OMe, Substance P methyl ester
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C64H99N17O14S
CAS Number:
Molecular Weight:
1362.64
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
assay
≥97% (HPLC)
UniProt accession no.
storage temp.
−20°C
SMILES string
COC(=O)C(CCSC)NC(=O)C(CC(C)C)NC(=O)CNC(=O)C(Cc1ccccc1)NC(=O)C(Cc2ccccc2)NC(=O)C(CCC(N)=O)NC(=O)C(CCC(N)=O)NC(=O)C3CCCN3C(=O)C(CCCCN)NC(=O)C4CCCN4C(=O)C(N)CCCNC(N)=N
Gene Information
human ... TAC1(6863)
Biochem/physiol Actions
Selective NK-1 receptor agonist
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
A M Yunker et al.
The American journal of physiology, 271(5 Pt 1), G904-G912 (1996-11-01)
Nitric oxide (NO) mediates neurogenic relaxations of gastrointestinal (GI) smooth muscle. NO synthase (NOS) inhibitors also alter neurogenic contractions, suggesting NO modulates excitatory neurotransmitter release. In circular muscle-myenteric plexus preparations, guanethidine and either scopolamine or CP-96,345, a neurokinin-1 (NK1) receptor
M Zhang et al.
Biopolymers, 33(12), 1901-1908 (1993-12-01)
High-resolution proton spectra at 500 MHz of two tachykinin peptides, substance P methyl ester (SPOMe) and [Nle10]-neurokinin A (4-10), have been obtained in dimethylsulfoxide (DMSO), and for SPOMe, also in 2,2,2-trifluoroethanol (TFE)/water mixtures. Complete chemical shift assignments for these peptides
D T Beattie et al.
British journal of pharmacology, 108(1), 146-149 (1993-01-01)
1. The effects of selective neurokinin agents on pial artery diameter, measured with an on-line image analyser, have been studied in anaesthetized guinea-pigs in order to characterize the neurokinin receptors present on pial arteries. 2. Perivascular injection of either substance
N D Fisher et al.
Neuroscience letters, 165(1-2), 84-88 (1994-01-03)
Using intracellular current clamp recording from motoneurones of the neonatal rat spinal cord in vitro, the action of tachykinin receptor agonists was investigated. Test drugs included the endogenously occurring neuropeptide substance P and synthetic compounds, such as substance P methylester
Y Tian et al.
Journal of neurochemistry, 67(3), 1191-1199 (1996-09-01)
Three chimeric receptors were constructed by exchanging exons between human neurokinin NK1 and NK3 receptor genes. The N-terminal sequences of these chimeric receptors are encoded by exon 1, exon 1-2, or exon 1-3 of the NK1 receptor gene, whereas the