Key Documents
S1945
Sigma-Aldrich
Anti-SF1 antibody produced in rabbit
affinity isolated antibody, ammonium sulfate suspension
Synonym(s):
Anti-Splicing Factor 1
Sign Into View Organizational & Contract Pricing
About This Item
MDL number:
UNSPSC Code:
12352203
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
ammonium sulfate suspension
mol wt
antigen 67.2 kDa
species reactivity
rodent, human
technique(s)
immunohistochemistry: suitable
western blot: suitable
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... SF1(7536)
mouse ... Sf1(22668)
rat ... Sf1(117855)
Immunogen
synthetic peptide corresponding to human SF1 (amino acids 10-25), conjugated to BSA.
Not finding the right product?  
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Miki Shitashige et al.
Gastroenterology, 132(3), 1039-1054 (2007-03-27)
beta-Catenin is the downstream effector of the Wnt signaling pathway and is involved in the process of colorectal carcinogenesis. However, it is still uncertain whether beta-catenin exerts its oncogenic function solely by coactivating the target genes of T-cell factor-4 (TCF4).