Key Documents
S0812
Sigma-Aldrich
Staphylococcal Enterotoxin B Fragment (150-161)
≥97% (HPLC)
Synonym(s):
SEB Fragment (150-161)
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C58H103N17O21
Molecular Weight:
1374.54
MDL number:
UNSPSC Code:
12352200
assay
≥97% (HPLC)
storage temp.
−20°C
Amino Acid Sequence
Thr-Asn-Lys-Lys-Lys-Val-Thr-Ala-Gln-Glu-Leu-Asp
Analysis Note
This peptide is not assayed for bioactivity.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)