About This Item
form
aqueous ethanol solution
extent of labeling
15-30 Ci/mmol
storage temp.
2-8°C
SMILES string
OC(=O)C1CCCN1
Packaging
Physical form
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.