Key Documents
P5444
Sigma-Aldrich
Polydeoxycytidylic acid sodium salt
Synonym(s):
Poly(dC)
Sign Into View Organizational & Contract Pricing
About This Item
CAS Number:
MDL number:
UNSPSC Code:
41106305
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Unit Definition
One unit will yield an A270 of 1.0 in 1.0 ml of 0.1 M NaCl, 0.02 M phosphate buffer, pH 7.0 (1 cm light path).
Preparation Note
Prepared by the action of terminal deoxynucleotidyl transferase on (dC)6.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Bollum, F.J.
Methods in Enzymology, 12(B), 591-591 (1968)