Key Documents
P4080
Sigma-Aldrich
pGlu-Gly-Arg p-nitroanilide hydrochloride
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C19H26N8O6 · HCl
CAS Number:
Molecular Weight:
498.92
MDL number:
UNSPSC Code:
12352204
storage temp.
−20°C
Application
substrate for urokinase
pictograms
Health hazardExclamation mark
signalword
Warning
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
K Huber et al.
Thrombosis and haemostasis, 47(3), 197-202 (1982-06-28)
Urokinase (UK) could be purified to apparent homogeneity starting from crude urine by sequential adsorption and elution of the enzyme to gelatine-Sepharose and agmatine-Sepharose followed by gel filtration on Sephadex G-150. The purified product exhibited characteristics of the high molecular
D D Bansal et al.
Endocrinology, 126(5), 2245-2251 (1990-05-01)
The characteristics and secretion of plasminogen activator (PA) were examined in fresh and cultured bovine parathyroid cells. Release of PA activity was maximal at low Ca concentrations ([Ca]) and suppressed at physiological [Ca]. PA secretion at high [Ca], like that
H H Ipsen et al.
Biochimica et biophysica acta, 613(2), 476-481 (1980-06-13)
The enzymic properties of urokinase (EC 3.4.21.31) were studied. The kinetic parameters of hydrolysis of 5-oxo-Pro-Gly-Arg-NA were determined in the pH range 5-9, at 25 degrees C and 37 degrees C. The reaction is affected by only one ionizing group
R Pannell et al.
Blood, 67(5), 1215-1223 (1986-05-01)
Highly purified pro-urokinase (pro-UK) or single-chain urokinase-type plasminogen activator (scu-PA) was treated with diisopropylfluorophosphate (1 mmol/L) to eliminate traces of two-chain UK activity. This preparation was found to retain a low activity against a chromogenic substrate (S2444), equivalent to 0.1%
Y Itoh et al.
Nihon Sanka Fujinka Gakkai zasshi, 35(1), 68-76 (1983-01-01)
Thrombolytic therapy with Urokinase (UK) has often been successful but it is very difficult to determine the effective dosage of UK. It is reported that after UK administration, plasmin fibrinolytic activity was immediately inhibited by alpha 2-Plasmin inhibitor (alpha 2-PI).