Key Documents
P3867
Sigma-Aldrich
Phenanthrene-9-14C
≥95% (Radiochemical Purity, HPLC), methanol solution
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C14H10
CAS Number:
Molecular Weight:
178.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
assay
≥95% (Radiochemical Purity, HPLC)
form
methanol solution
extent of labeling
40-60 mCi per mmol
mass shift
M+2
storage temp.
2-8°C
SMILES string
c1ccc2c(c1)c[14cH]c3ccccc23
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.