P3030
[Nle8,18, Tyr34]-PTH Fragment 3-34 amide bovine
≥97% (HPLC)
Synonym(s):
Ser-Glu-Ile-Gln-Phe-Nle-His-Asn-Leu-Gly-Lys-His-Leu-Ser-Ser-Nle-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Tyr-NH2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C177H279N53O48
CAS Number:
Molecular Weight:
3917.44
MDL number:
UNSPSC Code:
12352200
assay
≥97% (HPLC)
composition
Peptide content, ~70%
storage temp.
−20°C
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protein kinase-A and the effects of parathyroid hormone on phosphate uptake in opossum kidney cells.
K J Martin et al.
Endocrinology, 125(1), 295-301 (1989-07-01)
Current evidence indicates that signal transduction after receptor binding of PTH involves the stimulation of adenylate cyclase as well as stimulation of phosphoinositide metabolism. Recent studies, showing that PTH alters phosphate transport in opossum kidney cells at concentrations which do
T J Rosol et al.
Endocrinology, 122(5), 2098-2102 (1988-05-01)
In vitro bone resorption induced by tumor extract derived from a hypercalcemic canine adenocarcinoma was significantly inhibited by the PTH receptor antagonist, [Nle8,18,Tyr34]bovine (b) PTH-(3-34) amide. Dose-response curves were prepared for in vitro bone resorption induced in neonatal mouse calvaria
Identification of a renal receptor for parathyroid hormone by photoaffinity radiolabeling using a synthetic analogue.
M D Coltrera et al.
The Journal of biological chemistry, 256(20), 10555-10559 (1981-10-25)
J T Daugirdas et al.
Mineral and electrolyte metabolism, 13(1), 33-37 (1987-01-01)
We examined the vascular effects of bovine PTH(1-34) and of an analogue, Nle8,18Tyr34bPTH(3-34) amide, which inhibits adenylate cyclase activation by bPTH(1-34) in a number of in vitro systems. In the conscious dog. bPTH(1-34) injection produced dose-dependent hypotension associated with increased
H J Donahue et al.
The Journal of biological chemistry, 263(27), 13522-13527 (1988-09-25)
While the stimulatory effect of parathyroid hormone (PTH) on osteoblast-like cell adenylate cyclase is well known, the effect of PTH on cytosolic calcium ion ([Ca2+]i) mobilization is controversial, one group finding no effect but others reporting various increases. We investigated