P2303
Protein Kinase C fragment 530-558
≥97% (HPLC), powder
About This Item
assay
≥97% (HPLC)
form
powder
color
white
UniProt accession no.
Storage temp.
−20°C
SMILES string
CCC(C)C(NC(=O)C(CO)NC(=O)C(CCC(N)=O)NC(=O)C(Cc1ccccc1)NC(=O)C(CC(C)C)NC(=O)C(CCC(O)=O)NC(=O)C(CC(O)=O)NC(=O)C(CCC(O)=O)NC(=O)C(CC(O)=O)NC(=O)C(CCC(O)=O)NC(=O)CNC(=O)C(CCC(O)=O)NC(=O)C(Cc2ccccc2)NC(=O)C3CCCN3C(=O)C(C)NC(=O)C(CCC(N)=O)NC(=O)CNC(=O)C(C)NC(=O)C(CC(C)C)NC(=O)C(CCSC)NC(=O)C(CCC(O)=O)NC(=O)C(Cc4ccc(O)cc4)NC(=O)C(CC(C)C)NC(=O)C(N)CC(C)C)C(=O)NC(CCSC)C(=O)NC(CCC(O)=O)C(=O)NC(Cc5cnc[nH]5)C(=O)NC(CC(N)=O)C(=O)NC(C(C)C)C(N)=O
Gene Information
human ... PRKCA(5578)
Amino Acid Sequence
Biochem/physiol Actions
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?