Key Documents
P1368
Sigma-Aldrich
L-α-Phosphatidylinositol dipalmitoyl ammonium salt
≥98% (TLC)
Synonym(s):
1,2-Dihexadecanoyl-sn-glycero-3-phospho-(1-D-myo-inositol), DPPI
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C41H82NO13P
Molecular Weight:
828.06
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
assay
≥98% (TLC)
solubility
H2O: insoluble
organic solvents: soluble
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCCCCC(=O)OC[C@H](COP(O)(=O)O[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@@H](O)[C@H]1O)OC(=O)CCCCCCCCCCCCCCC
InChI
1S/C41H79O13P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(42)51-31-33(32-52-55(49,50)54-41-39(47)37(45)36(44)38(46)40(41)48)53-35(43)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h33,36-41,44-48H,3-32H2,1-2H3,(H,49,50)/t33-,36-,37-,38+,39-,40-,41-/m1/s1
InChI key
IBUKXRINTKQBRQ-KCKFLZCVSA-N
General description
Synthetic analog of the natural phosphatidyl inositols with C16:0 saturated fatty acids and the same inositol and diacyl glycerol stereochemistry as that of the natural compound.
Reconstitution
Can be dispersed by sonication to form an opaque but homogeneous preparation.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Jingjing Wang et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 96, 466-470 (2017-10-17)
Dipeptidyl peptidase I (DPPI), a lysosomal cysteine protease is derived from granule immune cells including mast cell, neutrophils, and toxicity T cells. DPPI can activate serine proteases by removal of dipeptides from N-termini of the pro-proteases, resulting in granule immune
Mingzhen Yang et al.
Journal of forensic sciences, 63(3), 745-751 (2017-08-24)
A method to determine postmortem interval (PMI) based on environmental temperature and the concentrations of vitreous humor (VH) molecules were explored. Rabbit carcasses were placed in a chamber at 5, 15, 25, or 35°C, and 80-100 μL of VH was
Yi Chu et al.
Cell biology international, 41(10), 1093-1102 (2017-07-01)
Dipeptidyl peptidase I (DPPI), a lysosomal cysteine protease, required for activation of serine proteases of granulocytes including mast cells (MCs), neutrophils (NPs) and others, which were found in synovial tissue of patients with rheumatoid arthritis (RA). But, the role of
Jingqiu He-Yang et al.
International immunopharmacology, 88, 106881-106881 (2020-08-17)
Necrotizing enterocolitis (NEC), a devastating infant disease characterized by severe intestinal necrosis, its pathogenesis is poorly understood, but appears to be multifactorial and highly associated with immaturity of gastrointestinal tract and immature innate-immune system. Breast-milk is effective strategy to protect
Masayuki Onishi et al.
Eukaryotic cell, 6(12), 2343-2353 (2007-10-24)
Sporulation of the fission yeast Schizosaccharomyces pombe is a developmental process that generates gametes and that includes the formation of spore envelope precursors called the forespore membranes. Assembly and development of forespore membranes require vesicular trafficking from other intracellular membrane