Key Documents
P1031
Sigma-Aldrich
Phorbol 13-butyrate
liquid
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C24H34O7
CAS Number:
Molecular Weight:
434.52
UNSPSC Code:
12352200
form
liquid
storage temp.
−20°C
SMILES string
CCCC(=O)O[C@@]12[C@H](O)[C@@H](C)[C@@]3(O)[C@@H](C=C(CO)C[C@@]4(O)[C@H]3C=C(C)C4=O)[C@@H]1C2(C)C
Packaging
Sealed ampule.
pictograms
Skull and crossbones
signalword
Danger
Hazard Classifications
Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.