Key Documents
P0871
Sigma-Aldrich
PETCM
≥98% (HPLC), solid
Synonym(s):
α-(Trichloromethyl)-4-pyridineethanol, 1-(4-Pyridyl)-2-hydroxy-3,3,3-trichloropropane
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C8H8NOCl3
Molecular Weight:
240.51
MDL number:
UNSPSC Code:
12352200
assay
≥98% (HPLC)
form
solid
solubility
DMSO: soluble 22 mg/mL
SMILES string
OC(Cc1ccncc1)C(Cl)(Cl)Cl
Biochem/physiol Actions
Apoptosis inducer; caspase 3 activator. Antagonizes prothymocin-α (ProT-α, oncoprotein) inhibition of apoptosome formation.
pictograms
Exclamation mark
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)