Key Documents
P0414
Sigma-Aldrich
Pyrophosphatase, Acid from tobacco
buffered aqueous glycerol solution, 5,000-10,000 units/mL
Synonym(s):
Acid pyrophosphatase
Sign Into View Organizational & Contract Pricing
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12352204
form
buffered aqueous glycerol solution
concentration
5,000-10,000 units/mL
shipped in
wet ice
storage temp.
−20°C
Unit Definition
One unit will release 1.0 nanomole of inorganic phosphate from 7-methyl-GTP in 30 min at pH 6.0 at 37 °C. One 7-methyl-GTP unit (pH 6.0) is equivalent to 15 ATP units (pH 5.0).
Physical form
Solution in 10 mM Tris HCl, pH 7.5, 10 mM NaCl, 0.1 mM EDTA, 1 mM DTT, 0.01% TRITON® X-100, 50% glycerol.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)