Key Documents
P0388
Sigma-Aldrich
Poly-L-hydroxyproline
mol wt 5,000-20,000
Sign Into View Organizational & Contract Pricing
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12352200
mol wt
5,000-20,000
storage temp.
−20°C
Analysis Note
Molecular weight based on viscosity. Also assayed by MALLS.
Other Notes
For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.
Choose from one of the most recent versions:
Certificates of Analysis (COA)