Key Documents
P0139
Sigma-Aldrich
S-[2-(4-Pyridyl)ethyl]-L-cysteine
suitable for amino acid analysis (Internal standard)
Synonym(s):
4-PEC, S-[β-(4-Pyridyl)ethyl]-L-cysteine
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C10H14N2O2S
CAS Number:
Molecular Weight:
226.30
Beilstein/REAXYS Number:
1536857
EC Number:
MDL number:
UNSPSC Code:
12352200
Quality Level
suitability
suitable for amino acid analysis (Internal standard)
Storage temp.
2-8°C
SMILES string
N[C@@H](CSCCc1ccncc1)C(O)=O
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
C S Fullmer
Analytical biochemistry, 142(2), 336-339 (1984-11-01)
A method is described for the facile identification of half-cystine residues in peptides from protein digests. Combined use of S-beta-(4-pyridylethylation) of cysteine residues with 4-vinyl pyridine and peptide mapping by reversed-phase high-performance liquid chromatography provides the basis for this procedure.
An internal standard for amino acid analyses: S-beta-(4-pyridylethyl)-L-cysteine.
J F Cavins et al.
Analytical biochemistry, 35(2), 489-493 (1969-11-01)