Key Documents
O6380
Sigma-Aldrich
[Thr4, Gly7]-Oxytocin
≥97% (HPLC)
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C39H61N11O12S2
CAS Number:
Molecular Weight:
940.10
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
assay
≥97% (HPLC)
storage temp.
−20°C
SMILES string
CCC(C)C1NC(=O)C(Cc2ccc(O)cc2)NC(=O)C(N)CSSCC(NC(=O)C(CC(N)=O)NC(=O)C(NC1=O)C(C)O)C(=O)NCC(=O)NC(CC(C)C)C(=O)NCC(N)=O
Amino Acid Sequence
Cys-Tyr-Ile-Thr-Asn-Cys-Gly-Leu-Gly-NH2 [Disulfide Bridge: 1-6]
Biochem/physiol Actions
Neuropeptide synthesized in the hypothalamus and involved in the control of lactation and parturition; selective ligand for central and peripheral oxytocin receptors.
pictograms
Exclamation mark
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Inhalation
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
The role of oxytocin: present concepts.
J S Jenkins et al.
Clinical endocrinology, 34(6), 515-525 (1991-06-01)
J Elands et al.
The American journal of physiology, 254(1 Pt 1), E31-E38 (1988-01-01)
Oxytocin receptors in rat hippocampal synaptic plasma membranes were compared with mammary gland and uterine oxytocin receptors. For this purpose, a highly specific oxytocic agonist [Thr4,Gly7]oxytocin was tritiated. We demonstrated that this ligand labels oxytocin receptors selectively. Scatchard analyses revealed
Neurotransmitter and neurohormonal regulation of oxytocin secretion in lactation.
W R Crowley et al.
Annals of the New York Academy of Sciences, 652, 286-302 (1992-06-12)
E Politowska et al.
Journal of receptor and signal transduction research, 22(1-4), 393-409 (2002-12-31)
The aim of the study was to computer-dock selected ligands to neurophyseal receptors in order to identify amino acid residues responsible for ligand-receptor interactions. To this aim, reliable oxytocin receptor (OTR) and arginine-vasopressin receptor (V1aR/V2R) models were built. The OTR-selective
Y H Jo et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 18(7), 2377-2386 (1998-04-16)
The functional characteristics of binding sites for the neuropeptide oxytocin (OT) detected by radioautography in laminae I and II of the dorsal horn (DH) and on cultured neonatal DH neurons were studied on the latter using perforated patch-clamp recordings. The