O1252
Octopine Dehydrogenase from queen scallop
lyophilized powder, ≥5 units/mg protein
Synonym(s):
N2-(1-Carboxyethyl)-L-arginine:NAD+ oxidoreductase (L-arginine forming)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12352204
form
lyophilized powder
specific activity
≥5 units/mg protein
composition
Protein, ≥60% Lowry
storage temp.
2-8°C
General description
Octopine dehydrogenase (OcDH) is from the adductor muscle of a scallop and it catalyses the NADH-dependent condensation of l-arginine and pyruvate to d-octopine, NAD(+) and water during escape swimming and subsequent recovery.
Application
Octopine dehydrogenase has been used in a study to assess metabolic and physiological responses in tissues of the long-lived bivalve Arctica islandica to oxygen deficiency. It has also been used in a study to investigate biochemical responses of the marine mussel Mytilus galloprovincialis to petrochemical environmental contamination.
Biochem/physiol Actions
Octopine Dehydrogenase activity can be inhibited by treatment with the dialdehyde, o-phthalaldehyde.
Unit Definition
One unit will form 1.0 μmole of octopine from 1.0 μmole of pyruvate and 1.0 μmole of L-arginine per min. at pH 6.6 at 25 °C.
Physical form
Contains Tris buffer salts
signalword
Danger
hcodes
pcodes
Hazard Classifications
Resp. Sens. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
B A Seibel et al.
The Biological bulletin, 198(2), 284-298 (2000-04-29)
The present study attempts to correlate the metabolism and locomotory behavior of 25 species of midwater Cephalopoda from California and Hawaii with the maximal activities of key metabolic enzymes in various locomotory muscle tissues. Citrate synthase (CS) and octopine dehydrogenase
J Jakowitsch et al.
The Plant journal : for cell and molecular biology, 17(2), 131-140 (1999-03-13)
One type of homology-dependent gene silencing in transgenic plants involves a silencing locus that is able to transcriptionally inactivate and methylate an unlinked target locus with which it shares sequence identity in promoter regions. In a manner resembling paramutation of
Rhonda C Foley et al.
FEBS letters, 563(1-3), 141-145 (2004-04-06)
TGA/OBF family members are bZIP transcription factors that bind to the octopine synthase (ocs) element, a plant promoter sequence that has been strongly linked to defence/stress responses. Intron-containing hairpin (ihp) constructs were used to generate Arabidopsis lines with reduced expression
Julia Strahl et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 158(4), 513-519 (2010-12-28)
In Arctica islandica, a long lifespan is associated with low metabolic activity, and with a pronounced tolerance to low environmental oxygen. In order to study metabolic and physiological responses to low oxygen conditions vs. no oxygen in mantle, gill, adductor
Ecophysiological influence on scaling of aerobic and anaerobic metabolism of pelagic gonatid squids.
Rui Rosa et al.
Physiological and biochemical zoology : PBZ, 82(5), 419-429 (2009-03-07)
We examined the oxygen consumption rates and activity levels of respiratory enzymes involved in the aerobic (citrate synthase [CS]) and anaerobic (octopine dehydrogenase [ODH]) metabolism of gonatid squids (Gonatus onyx and Gonatus pyrus) as a function of body size. The