Key Documents
N8888
Sigma-Aldrich
2-Nitrophenyl α-D-galacto­pyran­oside
>99% (TLC)
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C12H15NO8
CAS Number:
Molecular Weight:
301.25
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
assay
>99% (TLC)
storage temp.
−20°C
SMILES string
OCC1OC(Oc2ccccc2N(=O)=O)C(O)C(O)C1O
Looking for similar products? Visit Product Comparison Guide
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Sung Hun Yi et al.
New biotechnology, 28(6), 806-813 (2011-09-06)
After the complete gene of a β-galactosidase from human isolate Bifidobacterium breve B24 was isolated by PCR and overexpressed in E. coli, the recombinant β-galactosidase was purified to homogeneity and characterized for the glycoside transferase (GT) and glycoside hydrolase (GH)
Mireia Baeza et al.
Analytical chemistry, 82(3), 1006-1011 (2010-01-01)
Low-temperature cofired ceramics (LTCC) technology is a versatile fabrication technique used to construct microflow systems. It permits the integration of several unitary operations (pretreatment, separation, (bio)chemical reaction, and detection stage) of an analytical process in a modular or monolithic way.
Hiroyuki Honda et al.
The Journal of general and applied microbiology, 58(1), 11-17 (2012-03-28)
Lactobacillus gasseri ATCC33323(T) expresses four enzymes showing phospho-β-galactosidase activity (LacG1, LacG2, Pbg1 and Pbg2). We previously reported the purification and characterization of two phospho-β-galactosidases (Pbg1 and Pbg2) from Lactobacillus gasseri JCM1031 cultured in lactose medium. Here we aimed to characterize
Deepak Bhardwaj et al.
Plant physiology and biochemistry : PPB, 58, 1-5 (2012-07-04)
Climate change is a major concern especially in view of the increasing global population and food security. Plant scientists need to look for genetic tools whose appropriate usage can contribute to sustainable food availability. G-proteins have been identified as some
Kui Wang et al.
Applied microbiology and biotechnology, 88(1), 155-165 (2010-07-09)
A novel beta-galactosidase gene, zd410, was isolated by screening a soil metagenomic library. Sequence analysis revealed that zd410 encodes a protein of 672 amino acids with a predicted molecular weight of 78.6 kDa. The recombinant ZD410 was expressed and purified