Key Documents
N7646
Sigma-Aldrich
5-Nitrobenzimidazole nitrate salt
suitable for photographic applications, ≥95%
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C7H5N3O2 · HNO3
CAS Number:
Molecular Weight:
226.15
Beilstein/REAXYS Number:
8863868
EC Number:
MDL number:
UNSPSC Code:
12352100
assay
≥95%
mp
218-220 °C
suitability
suitable for photographic applications
SMILES string
O[N+]([O-])=O.[O-][N+](=O)c1ccc2[nH]cnc2c1
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)