Key Documents
N7521
Sigma-Aldrich
4-Nonylphenoxypolyglycidyl ether
suitable for photographic applications
Sign Into View Organizational & Contract Pricing
About This Item
MDL number:
UNSPSC Code:
12161900
description
non-ionic
suitability
suitable for photographic applications
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)