Key Documents
N7394
Sigma-Aldrich
Naphthalene-UL-14C
methanol solution
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C10H8
CAS Number:
Molecular Weight:
128.17
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
form
methanol solution
extent of labeling
40-60 mCi per mmol
storage temp.
2-8°C
SMILES string
c1ccc2ccccc2c1
Gene Information
Looking for similar products? Visit Product Comparison Guide
Packaging
sealed ampule
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Paloma Kachel Gusso-Choueri et al.
Environmental science and pollution research international, 22(22), 18047-18065 (2015-07-16)
Marine protected areas (MPAs) are vulnerable to many pressures, including pollution. However, environmental quality monitoring in these areas traditionally relies on only water chemistry and microbiological parameters. The goal of the current study was to investigate the role of a
Aliaksei Z Holik et al.
Respiratory research, 16, 67-67 (2015-06-07)
The lung is constantly exposed to environmental challenges and must rapidly respond to external insults. Mechanisms involved in the repair of the damaged lung involve expansion of different epithelial cells to repopulate the injured cellular compartment. However, factors regulating cell
Fabrice Gritti et al.
Journal of chromatography. A, 1352, 20-28 (2014-06-14)
The effect of the pressure on the dispersion of a low molecular weight compound along 0.508 and 1.016 mm i.d. × 50 cm long open circular tubes was investigated theoretically and experimentally. The theoretical predictions were based on the early
Martha L Bustos et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 23(3), 561-569 (2014-11-21)
The contribution of bone marrow cells (BMC) in lung repair is controversial. We previously reported a subpopulation of BMC that express Clara cell secretory protein (CCSP). To determine the contribution of endogenous CCSP(+) BMC to airway regeneration, we performed bone
Peta A Neale et al.
Journal of hazardous materials, 296, 192-200 (2015-05-02)
Tattooing is becoming increasingly popular, particularly amongst young people. However, tattoo inks contain a complex mixture of chemical impurities that may pose a long-term risk for human health. As a first step towards the risk assessment of these complex mixtures