Key Documents
N4279
Sigma-Aldrich
Neuropeptide Y Fragment 3-36 porcine
≥95% (HPLC)
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C176H271N53O54
CAS Number:
Molecular Weight:
3993.36
MDL number:
UNSPSC Code:
12352200
assay
≥95% (HPLC)
composition
Peptide content, ~60%
UniProt accession no.
storage temp.
−20°C
Gene Information
pig ... NPY(397304)
Amino Acid Sequence
Ser-Lys-Pro-Asp-Asn-Pro-Gly-Glu-Asp-Ala-Pro-Ala-Glu-Asp-Leu-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-NH2
Biochem/physiol Actions
Endogenous neuropeptide that selectively binds to Y2 and Y5 receptors.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
R Mentlein et al.
Regulatory peptides, 49(2), 133-144 (1993-12-10)
Neuropeptide Y, peptide YY and pancreatic polypeptide share an evolutionary conserved proline-rich N-terminal sequence, a structure generally known to be inert to the attack of common proteinases, but a potential target for specialized proline-specific aminopeptidases. Purified human dipeptidyl peptidase IV
K N Browning et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 12(1), 33-41 (2000-01-29)
Intracellular recordings were made from myenteric neurones of the guinea-pig descending colon. Neuropeptide Y (NPY) and related pancreatic polypeptides were applied by superfusion and the effects upon the amplitude of fast excitatory synaptic potentials (ESPs) and the ratio of paired
D Grandt et al.
Regulatory peptides, 67(1), 33-37 (1996-11-14)
Neuropeptide Y (NPY 1-36) binds to Y1 and Y2 receptors with similar affinity. No endogenous molecular form of NPY with selectivity for Y1 or Y2 receptors has been described so far. We report the presence of an endogenous fragment of