Key Documents
N3761
Sigma-Aldrich
1-Naphthyl red hydrochloride
Synonym(s):
4-(Phenylazo)-1-naphthalenamine hydrochloride
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
C6H5N=NC10H6NH2 · HCl
CAS Number:
Molecular Weight:
283.76
Colour Index Number:
11350
EC Number:
MDL number:
UNSPSC Code:
12171500
composition
Dye content, ~95%
SMILES string
Cl.Nc1ccc(N=Nc2ccccc2)c3ccccc13
InChI
1S/C16H13N3.ClH/c17-15-10-11-16(14-9-5-4-8-13(14)15)19-18-12-6-2-1-3-7-12;/h1-11H,17H2;1H
InChI key
CBIKBLARWHZKSA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)