All Photos(1)
Documents
N2161
Sigma-Aldrich
22-NBD-Cholesterol
Synonym(s):
(3,20S)-20-Methyl-21-((7-nitro-4-benzofurazanyl)-amino)-pregn-5-en-3-ol, 22-(N-(7-Nitrobenz-2-oxa-1,3-diazol-4-yl)amino)-23,24-bisnor-5-cholen-3-ol
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C28H38N4O4
Molecular Weight:
494.63
MDL number:
UNSPSC Code:
12352200
SMILES string
C[C@H](CNc1ccc([N+]([O-])=O)c2nonc12)[C@H]3CC[C@H]4[C@@H]5CC=C6C[C@@H](O)CC[C@]6(C)[C@H]5CC[C@]34C
Application
For investigation of lipid transport processes and lipid-protein interactions.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.