Key Documents
N1013
Sigma-Aldrich
Neoagarohexaose
≥95%
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C36H56O28
CAS Number:
Molecular Weight:
936.81
MDL number:
UNSPSC Code:
12352201
assay
≥95%
storage temp.
2-8°C
Other Notes
Mixed anomers
Preparation Note
Prepared enzymatically from agar.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
L M Morrice et al.
European journal of biochemistry, 137(1-2), 149-154 (1983-12-01)
Beta-Agarase I and II were characterised by their action on agar-type polysaccharides and oligosaccharides. Beta-Agarase I, an endo-enzyme, was specific for regions containing a minimum of one unsubstituted neoagarobiose unit [3,6-anhydro-alpha-L-galactopyranosyl-(1 leads to 3)-D-galactose], hydrolysing at the reducing side of