M9503
DL-Methionine DL-sulfoximine
Synonym(s):
DL-(3-Amino-3-carboxypropyl) methyl sulfoximine
About This Item
storage temp.
2-8°C
SMILES string
CS(=N)(=O)CCC(N)C(O)=O
InChI
1S/C5H12N2O3S/c1-11(7,10)3-2-4(6)5(8)9/h4,7H,2-3,6H2,1H3,(H,8,9)
InChI key
SXTAYKAGBXMACB-UHFFFAOYSA-N
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?