Key Documents
M9022
Sigma-Aldrich
Methyl 3-O-α-D-mannopyranosyl-α-D-mannopyranoside
>95% (HPLC)
Synonym(s):
α-D-Man-[1→3]-α-D-Man-1→OMe
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C13H24O11
CAS Number:
Molecular Weight:
356.32
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
assay
>95% (HPLC)
storage temp.
−20°C
SMILES string
COC1OC(CO)C(O)C(OC2OC(CO)C(O)C(O)C2O)C1O
Application
Useful in the study of UDP-N-acetylglucosamine: α-D-mannoside β-2-N-acetylglucosaminyltransferase.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
G J Vella et al.
Canadian journal of biochemistry and cell biology = Revue canadienne de biochimie et biologie cellulaire, 62(6), 409-417 (1984-06-01)
Twenty low molecular weight compounds were tested as substrates for UDP-GlcNAc:alpha-D-mannoside (GlcNAc to Man alpha 1-3) beta 2-N-acetylglucosaminyltransferase I (GlcNAc-transferase I) purified from bovine colostrum. This enzyme is at a key control point in the biosynthetic path leading to complex